Product Details

SNP ID
rs147044660
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:124539210 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATGGCGTTCTTGGCGTCGGGACC[C/T]TACCTGACCCATCAGCAAAAGGTGT
Phenotype
MIM: 601445
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NDUFB9 PubMed Links

Gene Details

Gene
NDUFB9
Gene Name
NADH:ubiquinone oxidoreductase subunit B9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278645.1 Intron NP_001265574.1
NM_001278646.1 Intron NP_001265575.1
NM_001311168.1 Intron NP_001298097.1
NM_005005.2 Intron NP_004996.1
Gene
TATDN1
Gene Name
TatD DNase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146160.1 Intron NP_001139632.1
NM_001317889.1 Intron NP_001304818.1
NM_001317890.1 Intron NP_001304819.1
NM_001317891.1 Intron NP_001304820.1
NM_032026.3 Intron NP_114415.1
XM_006716666.3 Intron XP_006716729.1
XM_006716667.3 Intron XP_006716730.1
XM_006716668.3 Intron XP_006716731.1
XM_006716669.3 Intron XP_006716732.1
XM_006716670.3 Intron XP_006716733.1
XM_006716671.3 Intron XP_006716734.1
XM_011517331.2 Intron XP_011515633.1
XM_017013895.1 Intron XP_016869384.1
XM_017013896.1 Intron XP_016869385.1
XM_017013897.1 Intron XP_016869386.1
XM_017013898.1 Intron XP_016869387.1
XM_017013899.1 Intron XP_016869388.1

View Full Product Details