Product Details

SNP ID
hCV174378066
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144390866 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGCCTGTGGTCAGAACAGCCCAG[C/G]CTACTTGGAGGTGATGTCTGCGTGT
Phenotype
MIM: 606027
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ADCK5 PubMed Links

Gene Details

Gene
ADCK5
Gene Name
aarF domain containing kinase 5
There are no transcripts associated with this gene.

Gene
CPSF1
Gene Name
cleavage and polyadenylation specific factor 1
There are no transcripts associated with this gene.

Gene
MIR1234
Gene Name
microRNA 1234
There are no transcripts associated with this gene.

Gene
MIR6849
Gene Name
microRNA 6849
There are no transcripts associated with this gene.

Gene
MIR939
Gene Name
microRNA 939
There are no transcripts associated with this gene.

View Full Product Details