Product Details

SNP ID
rs148963439
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:22118963 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCCTCCTCACCATCTGGAGAAAG[C/T]GGCGGATGCGCTGGGCGTCCTGTGC
Phenotype
MIM: 602302 MIM: 615791
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HR PubMed Links

Gene Details

Gene
HR
Gene Name
hair growth associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005144.4 3865 Missense Mutation CAC,CGC H1067R NP_005135.2
NM_018411.4 3865 Missense Mutation CAC,CGC H1067R NP_060881.2
XM_005273569.1 3865 Missense Mutation CAC,CGC H1068R XP_005273626.1
XM_006716367.1 3865 Missense Mutation CAC,CGC H1068R XP_006716430.1
Gene
NUDT18
Gene Name
nudix hydrolase 18
There are no transcripts associated with this gene.

View Full Product Details