Product Details
- SNP ID
-
rs149319759
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:6870817 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGATCTGTGGCCAAGGCCTGTGAGA[A/C]AGTTACCACCTGTAAGGAGGGAACA
- Phenotype
-
MIM: 602056
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DEFB1
PubMed Links
Gene Details
- Gene
- DEFB1
- Gene Name
- defensin beta 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005218.3 |
221 |
Missense Mutation |
TGT,TTT |
C24F |
NP_005209.1 |
View Full Product Details