Product Details

SNP ID
hCV174418996
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144317416 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAGGAGTGGAAGAGCCAGTAGAA[A/G]AAGATGAGCCAGATGAGGTGATTGG
Phenotype
MIM: 604900 MIM: 140580
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DGAT1 PubMed Links

Gene Details

Gene
DGAT1
Gene Name
diacylglycerol O-acyltransferase 1
There are no transcripts associated with this gene.

Gene
HSF1
Gene Name
heat shock transcription factor 1
There are no transcripts associated with this gene.

Gene
MIR6848
Gene Name
microRNA 6848
There are no transcripts associated with this gene.

View Full Product Details