Product Details

SNP ID
rs149824165
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:81660621 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATAATATGCATCTGCTCCGCCAG[C/T]TGCCACAAGGCACATATTAACAGCT
Phenotype
MIM: 602064
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IMPA1 PubMed Links

Gene Details

Gene
IMPA1
Gene Name
inositol monophosphatase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144878.1 941 Missense Mutation ACT,GCT T264A NP_001138350.1
NM_001144879.1 941 Silent Mutation CAA,CAG Q168Q NP_001138351.1
NM_005536.3 941 Missense Mutation ACT,GCT T205A NP_005527.1

View Full Product Details