Product Details

SNP ID
rs151081625
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144360027 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGTGCCCGCCAGCCCCCATCAAC[A/G]GCACCCCTGGCCCCCCGCTCGACTT
Phenotype
MIM: 609076 MIM: 607882
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBXL6 PubMed Links

Gene Details

Gene
FBXL6
Gene Name
F-box and leucine rich repeat protein 6
There are no transcripts associated with this gene.

Gene
LOC101928902
Gene Name
uncharacterized LOC101928902
There are no transcripts associated with this gene.

Gene
SLC52A2
Gene Name
solute carrier family 52 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001253815.1 1096 Missense Mutation AGC,GGC S179G NP_001240744.1
NM_001253816.1 1096 Missense Mutation AGC,GGC S179G NP_001240745.1
NM_024531.4 1096 Missense Mutation AGC,GGC S179G NP_078807.1
XM_006716658.2 1096 Missense Mutation AGC,GGC S179G XP_006716721.1
XM_006716659.2 1096 Missense Mutation AGC,GGC S179G XP_006716722.1
XM_006716660.2 1096 Missense Mutation AGC,GGC S179G XP_006716723.1
XM_011517300.2 1096 Missense Mutation AGC,GGC S91G XP_011515602.1
XM_017013819.1 1096 Missense Mutation AGC,GGC S179G XP_016869308.1
XM_017013820.1 1096 Missense Mutation AGC,GGC S179G XP_016869309.1
XM_017013821.1 1096 Intron XP_016869310.1
XM_017013822.1 1096 Intron XP_016869311.1
Gene
TMEM249
Gene Name
transmembrane protein 249
There are no transcripts associated with this gene.

View Full Product Details