Product Details

SNP ID
rs151193447
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:81658736 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTACTTCAAACCTGAAGCAAAAG[A/T]ACAGAATGCTGAGTTCATTACTTTA
Phenotype
MIM: 602064
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
IMPA1 PubMed Links

Gene Details

Gene
IMPA1
Gene Name
inositol monophosphatase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144878.1 1777 UTR 3 NP_001138350.1
NM_001144879.1 1777 UTR 3 NP_001138351.1
NM_005536.3 1777 UTR 3 NP_005527.1

View Full Product Details