Product Details

SNP ID
rs138324405
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:122661945 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGACATTTGCTATGTGACAGTCA[G/T]TATCCCTAAGATGCTGGTGAACTTC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OR1B1 PubMed Links

Gene Details

Gene
OR1B1
Gene Name
olfactory receptor family 1 subfamily B member 1 (gene/pseudogene)
There are no transcripts associated with this gene.

Gene
OR1L1
Gene Name
olfactory receptor family 1 subfamily L member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005236.3 230 Missense Mutation AGT,ATT S77I NP_001005236.3

View Full Product Details