Product Details

SNP ID
rs138476552
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:114790529 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGAGATGTCACTGACGTTCACCA[G/T]TAGCTTGTCCCCTTCTTGCAAGGAG
Phenotype
MIM: 604052
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TNFSF15 PubMed Links

Gene Details

Gene
TNFSF15
Gene Name
tumor necrosis factor superfamily member 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204344.1 795 Missense Mutation ATG,CTG M168L NP_001191273.1
NM_005118.3 795 Missense Mutation ATG,CTG M227L NP_005109.2

View Full Product Details