Product Details

SNP ID
rs138586605
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137460694 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCTGCGTATGCATCCCTGGGGAC[A/G]TCCAGCAGCTCCTCCTCGTACTCCG
Phenotype
MIM: 608137 MIM: 612122
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NSMF PubMed Links

Gene Details

Gene
NSMF
Gene Name
NMDA receptor synaptonuclear signaling and neuronal migration factor
There are no transcripts associated with this gene.

Gene
PNPLA7
Gene Name
patatin like phospholipase domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098537.2 4257 Silent Mutation GAC,GAT D1295D NP_001092007.2
NM_152286.4 4257 Silent Mutation GAC,GAT D1270D NP_689499.4
XM_006717102.1 4257 Silent Mutation GAC,GAT D1296D XP_006717165.1
XM_006717104.3 4257 Silent Mutation GAC,GAT D1262D XP_006717167.1
XM_011518664.2 4257 Intron XP_011516966.1
XM_017014709.1 4257 Silent Mutation GAC,GAT D1262D XP_016870198.1

View Full Product Details