Product Details

SNP ID
rs138717272
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:127944635 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTACCTGGTGGCAAGCTGGATGCC[A/G]CTCAGCGTGGCAGAGCCATCGCGGC
Phenotype
MIM: 603564 MIM: 610891
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DPM2 PubMed Links

Gene Details

Gene
DPM2
Gene Name
dolichyl-phosphate mannosyltransferase polypeptide 2, regulatory subunit
There are no transcripts associated with this gene.

Gene
FAM102A
Gene Name
family with sequence similarity 102 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001035254.2 967 Silent Mutation AGC,AGT S363S NP_001030331.1
NM_203305.2 967 Silent Mutation AGC,AGT S221S NP_976050.1

View Full Product Details