Product Details

SNP ID
rs139063852
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:2717961 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGAAGGACGACCTGGCAGAAGA[C/G]GACCAGCAGGCAGGGGAGGTCACCA
Phenotype
MIM: 607604
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KCNV2 PubMed Links

Gene Details

Gene
KCNV2
Gene Name
potassium voltage-gated channel modifier subfamily V member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_133497.3 436 Missense Mutation GAC,GAG D74E NP_598004.1

View Full Product Details