Product Details

SNP ID
rs139122611
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:120799459 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGGTTTTCTGGGAAAAGGCACAT[G/T]AGATACAACAGATGTTGCTATAAGT
Phenotype
MIM: 609071 MIM: 604452
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FBXW2 PubMed Links

Gene Details

Gene
FBXW2
Gene Name
F-box and WD repeat domain containing 2
There are no transcripts associated with this gene.

Gene
LOC100288842
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 5 pseudogene
There are no transcripts associated with this gene.

Gene
PSMD5
Gene Name
proteasome 26S subunit, non-ATPase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270427.1 Intron NP_001257356.1
NM_005047.3 Intron NP_005038.1
XM_011518866.2 Intron XP_011517168.1
XM_017014927.1 Intron XP_016870416.1
XM_017014928.1 Intron XP_016870417.1

View Full Product Details