Product Details

SNP ID
rs139667057
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:124779410 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCTGGGAGGGATATTTGGTGTT[A/G]GGCATCGTCTGTGCAGAGAGCTGTA
Phenotype
MIM: 602778 MIM: 615899
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NR6A1 PubMed Links

Gene Details

Gene
NR6A1
Gene Name
nuclear receptor subfamily 6 group A member 1
There are no transcripts associated with this gene.

Gene
OLFML2A
Gene Name
olfactomedin like 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282715.1 Intron NP_001269644.1
NM_182487.3 Intron NP_872293.2
XM_005251760.4 Intron XP_005251817.1
XM_006716989.2 Intron XP_006717052.1

View Full Product Details