Product Details

SNP ID
rs139668393
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:133379081 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGGCCTTGGGGGTGAACCTGGTG[A/G]TGGAGGAAATGGAAACCAAAGTCAA
Phenotype
MIM: 185660
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
STKLD1 PubMed Links

Gene Details

Gene
STKLD1
Gene Name
serine/threonine kinase like domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153710.4 194 Missense Mutation ATG,GTG M45V NP_714921.4
Gene
SURF4
Gene Name
surfeit 4
There are no transcripts associated with this gene.

View Full Product Details