Product Details

SNP ID
rs139727695
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:120858187 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAAGCCCCCTCTGCACTGCTGTC[C/T]GAGGGGCAGCGTCCATCCAGCTCAG
Phenotype
MIM: 609740
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PHF19 PubMed Links

Gene Details

Gene
PHF19
Gene Name
PHD finger protein 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009936.2 1624 Intron NP_001009936.1
NM_001286840.1 1624 Silent Mutation TCA,TCG S519S NP_001273769.1
NM_001286842.1 1624 Silent Mutation TCA,TCG S291S NP_001273771.1
NM_001286843.1 1624 Intron NP_001273772.1
NM_015651.2 1624 Silent Mutation TCA,TCG S500S NP_056466.1
XM_005251906.2 1624 Silent Mutation TCA,TCG S500S XP_005251963.1
XM_011518509.2 1624 Silent Mutation TCA,TCG S500S XP_011516811.1
XM_011518511.2 1624 UTR 3 XP_011516813.1
XM_011518515.2 1624 Intron XP_011516817.1
XM_011518516.2 1624 Intron XP_011516818.1
XM_017014612.1 1624 Silent Mutation TCA,TCG S500S XP_016870101.1
XM_017014613.1 1624 Intron XP_016870102.1
Gene
PSMD5-AS1
Gene Name
PSMD5 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details