Product Details

SNP ID
rs140719684
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137460466 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGGGGTGTCGATGCCGCAGTGAG[A/G]ACTCGTCCTCCTGCAAGCAGACCGC
Phenotype
MIM: 608137 MIM: 612122
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NSMF PubMed Links

Gene Details

Gene
NSMF
Gene Name
NMDA receptor synaptonuclear signaling and neuronal migration factor
There are no transcripts associated with this gene.

Gene
PNPLA7
Gene Name
patatin like phospholipase domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098537.2 4328 Missense Mutation TCC,TTC S1319F NP_001092007.2
NM_152286.4 4328 Missense Mutation TCC,TTC S1294F NP_689499.4
XM_006717102.1 4328 Missense Mutation TCC,TTC S1320F XP_006717165.1
XM_006717104.3 4328 Missense Mutation TCC,TTC S1286F XP_006717167.1
XM_011518664.2 4328 Intron XP_011516966.1
XM_017014709.1 4328 Missense Mutation TCC,TTC S1286F XP_016870198.1

View Full Product Details