Product Details

SNP ID
rs141127395
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:37888515 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATCTTGTTTTGAAGATGTTAGTAT[C/T]GGTGGCCTCTTTTCATGAGCTTCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC25A51 PubMed Links

Gene Details

Gene
SLC25A51
Gene Name
solute carrier family 25 member 51
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033412.3 486 Silent Mutation NP_219480.1

View Full Product Details