Product Details

SNP ID
rs141391682
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:126615538 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACCACCAGCTGCTACTTCCGGGAT[A/C]GGAAACTGTACTGCAAACAAGACTA
Phenotype
MIM: 602575
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LMX1B PubMed Links

Gene Details

Gene
LMX1B
Gene Name
LIM homeobox transcription factor 1 beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174146.1 302 Silent Mutation AGG,CGG R99R NP_001167617.1
NM_001174147.1 302 Silent Mutation AGG,CGG R99R NP_001167618.1
NM_002316.3 302 Silent Mutation AGG,CGG R99R NP_002307.2

View Full Product Details