Product Details

SNP ID
rs141454067
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:114060965 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCAGTACTCTCTGCACTCCTTCTC[C/T]GAGTAGAACTTGTTCCCGTTGCCCT
Phenotype
MIM: 176870
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AMBP PubMed Links

Gene Details

Gene
AMBP
Gene Name
alpha-1-microglobulin/bikunin precursor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001633.3 1250 Silent Mutation TCC,TCT S329S NP_001624.1
Gene
ZNF618
Gene Name
zinc finger protein 618
There are no transcripts associated with this gene.

View Full Product Details