Product Details

SNP ID
rs142348267
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:104694563 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCATGACTGAATTCTTTCTGGTG[A/G]GGCTTTCCCAATATCCAGAGCTCCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR13D1 PubMed Links

Gene Details

Gene
OR13D1
Gene Name
olfactory receptor family 13 subfamily D member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004484.1 142 Missense Mutation AGG,GGG R48G NP_001004484.1

View Full Product Details