Product Details

SNP ID
rs142439188
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:120858229 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCTCAGCTGCCCACCGCTTTGC[C/T]CGCAGGGGCATGTATGCCTTGGCTG
Phenotype
MIM: 609740
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PHF19 PubMed Links

Gene Details

Gene
PHF19
Gene Name
PHD finger protein 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009936.2 1582 Intron NP_001009936.1
NM_001286840.1 1582 Silent Mutation CGA,CGG R505R NP_001273769.1
NM_001286842.1 1582 Silent Mutation CGA,CGG R277R NP_001273771.1
NM_001286843.1 1582 Intron NP_001273772.1
NM_015651.2 1582 Silent Mutation CGA,CGG R486R NP_056466.1
XM_005251906.2 1582 Silent Mutation CGA,CGG R486R XP_005251963.1
XM_011518509.2 1582 Silent Mutation CGA,CGG R486R XP_011516811.1
XM_011518511.2 1582 UTR 3 XP_011516813.1
XM_011518515.2 1582 Intron XP_011516817.1
XM_011518516.2 1582 Intron XP_011516818.1
XM_017014612.1 1582 Silent Mutation CGA,CGG R486R XP_016870101.1
XM_017014613.1 1582 Intron XP_016870102.1
Gene
PSMD5-AS1
Gene Name
PSMD5 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details