Product Details

SNP ID
rs142573111
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35615992 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTCAGTCTGTTCTCCATGTTGCT[C/G]AGCTTCTGCTCCAAGGCCCTCCTCT
Phenotype
MIM: 107272 MIM: 601782
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CD72 PubMed Links

Gene Details

Gene
CD72
Gene Name
CD72 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001782.2 1136 Silent Mutation CTC,CTG L213L NP_001773.1
XM_006716893.2 1136 Silent Mutation CTC,CTG L213L XP_006716956.1
XM_006716894.3 1136 Silent Mutation CTC,CTG L209L XP_006716957.1
XM_011518074.2 1136 Silent Mutation CTC,CTG L209L XP_011516376.1
XM_017015351.1 1136 Silent Mutation CTC,CTG L209L XP_016870840.1
Gene
MIR4667
Gene Name
microRNA 4667
There are no transcripts associated with this gene.

Gene
TESK1
Gene Name
testis-specific kinase 1
There are no transcripts associated with this gene.

View Full Product Details