Product Details

SNP ID
rs142589658
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137459999 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAACATCACACGGCAGCATCAGG[G/T]CTCCCACACCTCACAGGGCAGCAGG
Phenotype
MIM: 608137 MIM: 612122
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MIR7114 PubMed Links

Gene Details

Gene
MIR7114
Gene Name
microRNA 7114
There are no transcripts associated with this gene.

Gene
NSMF
Gene Name
NMDA receptor synaptonuclear signaling and neuronal migration factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130969.1 4795 Intron NP_001124441.1
NM_001130970.1 4795 Intron NP_001124442.1
NM_001130971.1 4795 Intron NP_001124443.1
NM_001178064.1 4795 Intron NP_001171535.1
NM_015537.4 4795 Intron NP_056352.3
XM_005266061.4 4795 Intron XP_005266118.1
XM_005266062.4 4795 Intron XP_005266119.1
XM_011518496.2 4795 Intron XP_011516798.1
XM_011518497.2 4795 Intron XP_011516799.1
XM_017014597.1 4795 Intron XP_016870086.1
Gene
PNPLA7
Gene Name
patatin like phospholipase domain containing 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098537.2 4795 UTR 3 NP_001092007.2
NM_152286.4 4795 UTR 3 NP_689499.4
XM_006717102.1 4795 UTR 3 XP_006717165.1
XM_006717104.3 4795 UTR 3 XP_006717167.1
XM_011518664.2 4795 Intron XP_011516966.1
XM_017014709.1 4795 UTR 3 XP_016870198.1

View Full Product Details