Product Details

SNP ID
rs142664086
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128345307 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCACAGATACCCACTGGACCTTCC[A/G]CCAGCTGGATGAGTACTCAAGCAGT
Phenotype
MIM: 604194
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC27A4 PubMed Links

Gene Details

Gene
SLC27A4
Gene Name
solute carrier family 27 member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005094.3 472 Missense Mutation CAC,CGC H105R NP_005085.2
XM_017014222.1 472 Missense Mutation CAC,CGC H105R XP_016869711.1

View Full Product Details