Product Details

SNP ID
rs142727320
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:37780615 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATCTTCTGAGTATTTAAATGGGG[A/G]AGGTTCACCTGAAAAAACCCATAGT
Phenotype
MIM: 606489
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EXOSC3 PubMed Links

Gene Details

Gene
EXOSC3
Gene Name
exosome component 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002269.2 940 UTR 3 NP_001002269.1
NM_016042.3 940 UTR 3 NP_057126.2
Gene
LOC105376037
Gene Name
uncharacterized LOC105376037
There are no transcripts associated with this gene.

Gene
TRMT10B
Gene Name
tRNA methyltransferase 10B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286950.1 940 Intron NP_001273879.1
NM_001286951.1 940 Intron NP_001273880.1
NM_001286952.1 940 Intron NP_001273881.1
NM_001286953.1 940 Intron NP_001273882.1
NM_001286954.1 940 Intron NP_001273883.1
NM_144964.3 940 Intron NP_659401.2
XM_005251375.2 940 Intron XP_005251432.1
XM_005251379.2 940 Intron XP_005251436.1
XM_011517735.2 940 Intron XP_011516037.1
XM_011517736.2 940 Intron XP_011516038.1
XM_011517738.2 940 Intron XP_011516040.1
XM_011517739.2 940 Intron XP_011516041.1
XM_011517742.1 940 Intron XP_011516044.1
XM_011517743.2 940 Intron XP_011516045.1
XM_017014313.1 940 Intron XP_016869802.1

View Full Product Details