Product Details

SNP ID
rs142823140
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:122661785 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGACTCTCCTCTCGACCTGAGGAT[C/T]AGAAGCCGCTCTTTGCTGTGTTCCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR1B1 PubMed Links

Gene Details

Gene
OR1B1
Gene Name
olfactory receptor family 1 subfamily B member 1 (gene/pseudogene)
There are no transcripts associated with this gene.

Gene
OR1L1
Gene Name
olfactory receptor family 1 subfamily L member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005236.3 70 Nonsense Mutation CAG,TAG Q24* NP_001005236.3

View Full Product Details