Product Details

SNP ID
rs142996260
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35606936 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCTGGACATTGCCCGAGGCCTG[C/T]GGTACCTGCACTCCAAAGGTGTATT
Phenotype
MIM: 107272 MIM: 601782
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD72 PubMed Links

Gene Details

Gene
CD72
Gene Name
CD72 molecule
There are no transcripts associated with this gene.

Gene
MIR4667
Gene Name
microRNA 4667
There are no transcripts associated with this gene.

Gene
TESK1
Gene Name
testis-specific kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318230.1 826 Silent Mutation TGC,TGT C31C NP_001305159.1
NM_006285.2 826 Missense Mutation CGG,TGG R164W NP_006276.2

View Full Product Details