Product Details

SNP ID
rs143331649
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34382826 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTGTAAGGTTCCATCCTCCATGT[A/G]TTGTACCAGTCACTGCCCAGGTAGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C9orf24 PubMed Links

Gene Details

Gene
C9orf24
Gene Name
chromosome 9 open reading frame 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252195.1 542 Intron NP_001239124.1
NM_032596.3 542 Silent Mutation AAC,AAT N108N NP_115985.2
NM_147168.1 542 Intron NP_671697.1
NM_147169.2 542 Intron NP_671698.1
XM_005251616.4 542 Silent Mutation AAC,AAT N108N XP_005251673.1
XM_005251618.4 542 Intron XP_005251675.1
XM_011518055.2 542 Intron XP_011516357.1
Gene
KIAA1161
Gene Name
KIAA1161
There are no transcripts associated with this gene.

View Full Product Details