Product Details
- SNP ID
-
rs143424189
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:76388603 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCTCAGGTAGCCAACAATGGCCAC[A/G]TTGAGGATTTCCCCATAGAAGTCCT
- Phenotype
-
MIM: 613010
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RFK
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10781358] are located under a probe and SNP(s) [rs2501925] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RFK
- Gene Name
- riboflavin kinase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018339.5 |
645 |
Silent Mutation |
AAC,AAT |
N96N |
NP_060809.3 |
- Gene
- RPSAP9
- Gene Name
- ribosomal protein SA pseudogene 9
There are no transcripts associated with this gene.
View Full Product Details