Product Details

SNP ID
rs143477600
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:38395950 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCACCGGGGAGGTCATTGGGCAC[A/G]TGGCTGAAGGTGACCGGGCTGATGT
Phenotype
MIM: 100670
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALDH1B1 PubMed Links

Gene Details

Gene
ALDH1B1
Gene Name
aldehyde dehydrogenase 1 family member B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000692.4 355 Missense Mutation ATG,GTG M68V NP_000683.3
XM_011517802.2 355 Missense Mutation ATG,GTG M68V XP_011516104.1

View Full Product Details