Product Details

SNP ID
rs143630506
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:96641395 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTGAATATAATGGTCTTGTGTTG[C/T]TCTCCTGTCCCCTTATATACTGTCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AAED1 PubMed Links

Gene Details

Gene
AAED1
Gene Name
AhpC/TSA antioxidant enzyme domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153698.1 1081 UTR 3 NP_714542.1
XM_005251783.3 1081 UTR 3 XP_005251840.1
XM_005251784.4 1081 Intron XP_005251841.1

View Full Product Details