Product Details

SNP ID
rs143854005
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128946382 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCACCACCAGGCAGGCCAAGGTGG[C/T]CAGCAGAGACCAATAGGCTAGGAGG
Phenotype
MIM: 610746 MIM: 615587
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DOLK PubMed Links

Gene Details

Gene
DOLK
Gene Name
dolichol kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014908.3 1352 Missense Mutation ACC,GCC T308A NP_055723.1
Gene
NUP188
Gene Name
nucleoporin 188
There are no transcripts associated with this gene.

Gene
PHYHD1
Gene Name
phytanoyl-CoA dioxygenase domain containing 1
There are no transcripts associated with this gene.

View Full Product Details