Product Details

SNP ID
rs144191733
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:110366105 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTCCTATGTGGTGCCAGTGGCAC[A/G]GATGAAAGATCATGAGTCCTCTCTA
Phenotype
MIM: 611691
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SVEP1 PubMed Links

Gene Details

Gene
SVEP1
Gene Name
sushi, von Willebrand factor type A, EGF and pentraxin domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153366.3 11487 UTR 3 NP_699197.3

View Full Product Details