Product Details
- SNP ID
-
rs144617096
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:70387806 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGCGTTGGCCAGCGCCTTTTTCGA[C/T]CGCTTGATCATGCTGGGGTGGAACT
- Phenotype
-
MIM: 602902
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KLF9
PubMed Links
Gene Details
- Gene
- KLF9
- Gene Name
- Kruppel like factor 9
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001206.2 |
1999 |
Silent Mutation |
CGA,CGG |
R235R |
NP_001197.1 |
View Full Product Details