Product Details

SNP ID
rs145390047
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:135547909 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCATATACCTGCAGGAGCTGCCC[A/G]GGACGGACGACTACGTCTTTTACTG
Phenotype
MIM: 164320
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OBP2A PubMed Links

Gene Details

Gene
OBP2A
Gene Name
odorant binding protein 2A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001293189.1 Intron NP_001280118.1
NM_001293193.1 Intron NP_001280122.1
NM_014582.2 Intron NP_055397.1
XM_017014656.1 Intron XP_016870145.1

View Full Product Details