Product Details

SNP ID
rs145405879
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:104504390 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCTGATCACCTTGCTGGGCAACA[C/T]TTTTCTGATCTCCATCACCATTCTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR13F1 PubMed Links

Gene Details

Gene
OR13F1
Gene Name
olfactory receptor family 13 subfamily F member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004485.1 128 Missense Mutation ACT,ATT T43I NP_001004485.1

View Full Product Details