Product Details

SNP ID
rs145408790
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:113313263 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTTTATTGGTAACAGAACCCTAC[C/T]AACTCATCAGAGCCCCACTATTTTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNF183 PubMed Links

Gene Details

Gene
RNF183
Gene Name
ring finger protein 183
There are no transcripts associated with this gene.

Gene
WDR31
Gene Name
WD repeat domain 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006615.2 4892 UTR 3 NP_001006616.1
NM_001012361.3 4892 UTR 3 NP_001012361.1
NM_145241.4 4892 UTR 3 NP_660284.1
XM_011518194.2 4892 Intron XP_011516496.1

View Full Product Details