Product Details

SNP ID
rs145798184
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:74952412 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCACGGCGCTTGCTGGGCGAAGCC[A/G]AGGGCTCTGCCATGGTCCCTGCGTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C9orf40 PubMed Links

Gene Details

Gene
C9orf40
Gene Name
chromosome 9 open reading frame 40
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017998.2 475 Missense Mutation TCG,TTG S67L NP_060468.2
Gene
C9orf41-AS1
Gene Name
C9orf41 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details