Product Details

SNP ID
rs146186151
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128792141 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCAGAGCATCGATGAATGAGAGC[C/T]GAAGAAACCAAATTCATTCATTCAG
Phenotype
MIM: 616218
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TBC1D13 PubMed Links

Gene Details

Gene
TBC1D13
Gene Name
TBC1 domain family member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286772.1 Intron NP_001273701.1
NM_018201.4 Intron NP_060671.3
XM_005252060.2 Intron XP_005252117.1

View Full Product Details