Product Details

SNP ID
rs146255999
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34689833 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACGGTCATAGGTTAACTGCTGCGGC[A/G]CTTCATCTAGAGGAGGAAGGAGAGG
Phenotype
MIM: 602227
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCL19 PubMed Links

Gene Details

Gene
CCL19
Gene Name
C-C motif chemokine ligand 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006274.2 421 Missense Mutation CGC,TGC R95C NP_006265.1

View Full Product Details