Product Details

SNP ID
rs146272196
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:133389560 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGTGCTGGGCCAGGTGCTGGACG[C/T]GCTGGAATACCTGCACCATTTGGAC
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
STKLD1 PubMed Links

Gene Details

Gene
STKLD1
Gene Name
serine/threonine kinase like domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153710.4 492 Missense Mutation GCG,GTG A144V NP_714921.4

View Full Product Details