Product Details
- SNP ID
-
rs146295355
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:125444594 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTAGATACGTGAGTTTAAATATTGC[A/G]TGACCTGCAGAGACAGAAAACTGTG
- Phenotype
-
MIM: 610558
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MAPKAP1
PubMed Links
Gene Details
- Gene
- MAPKAP1
- Gene Name
- mitogen-activated protein kinase associated protein 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001006617.1 |
1575 |
Silent Mutation |
CAC,CAT |
H450H |
NP_001006618.1 |
NM_001006618.1 |
1575 |
Intron |
|
|
NP_001006619.1 |
NM_001006619.1 |
1575 |
Silent Mutation |
CAC,CAT |
H403H |
NP_001006620.1 |
NM_001006620.1 |
1575 |
Silent Mutation |
CAC,CAT |
H258H |
NP_001006621.1 |
NM_001006621.1 |
1575 |
Silent Mutation |
CAC,CAT |
H258H |
NP_001006622.1 |
NM_024117.3 |
1575 |
Silent Mutation |
CAC,CAT |
H414H |
NP_077022.1 |
XM_011519005.1 |
1575 |
Silent Mutation |
CAC,CAT |
H497H |
XP_011517307.1 |
XM_011519006.2 |
1575 |
Silent Mutation |
CAC,CAT |
H461H |
XP_011517308.1 |
XM_011519007.1 |
1575 |
Silent Mutation |
CAC,CAT |
H450H |
XP_011517309.1 |
XM_011519008.1 |
1575 |
Intron |
|
|
XP_011517310.1 |
XM_011519009.1 |
1575 |
Intron |
|
|
XP_011517311.1 |
XM_011519010.2 |
1575 |
Silent Mutation |
CAC,CAT |
H258H |
XP_011517312.1 |
XM_017015126.1 |
1575 |
Silent Mutation |
CAC,CAT |
H450H |
XP_016870615.1 |
XM_017015127.1 |
1575 |
Silent Mutation |
CAC,CAT |
H414H |
XP_016870616.1 |
View Full Product Details