Product Details
- SNP ID
-
rs146402491
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:128835400 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTCCAGGCAAGTCAGGCATCTTCC[C/T]CTCTGGGAGACAGAGGCCACACTGA
- Phenotype
-
MIM: 600547
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KYAT1
PubMed Links
Gene Details
- Gene
- KYAT1
- Gene Name
- kynurenine aminotransferase 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001122671.1 |
1238 |
Missense Mutation |
AGG,GGG |
R349G |
NP_001116143.1 |
NM_001122672.1 |
1238 |
Missense Mutation |
AGG,GGG |
R299G |
NP_001116144.1 |
NM_001287390.1 |
1238 |
Missense Mutation |
AGG,GGG |
R443G |
NP_001274319.1 |
NM_004059.4 |
1238 |
Missense Mutation |
AGG,GGG |
R349G |
NP_004050.3 |
XM_006717311.3 |
1238 |
Missense Mutation |
AGG,GGG |
R383G |
XP_006717374.1 |
XM_011519165.2 |
1238 |
Missense Mutation |
AGG,GGG |
R442G |
XP_011517467.1 |
XM_011519167.2 |
1238 |
Missense Mutation |
AGG,GGG |
R396G |
XP_011517469.2 |
XM_011519168.1 |
1238 |
Missense Mutation |
AGG,GGG |
R349G |
XP_011517470.1 |
XM_011519169.1 |
1238 |
Missense Mutation |
AGG,GGG |
R349G |
XP_011517471.1 |
XM_011519170.2 |
1238 |
Missense Mutation |
AGG,GGG |
R349G |
XP_011517472.1 |
XM_017015260.1 |
1238 |
Missense Mutation |
AGG,GGG |
R456G |
XP_016870749.1 |
XM_017015261.1 |
1238 |
Missense Mutation |
AGG,GGG |
R443G |
XP_016870750.1 |
XM_017015262.1 |
1238 |
Missense Mutation |
AGG,GGG |
R443G |
XP_016870751.1 |
XM_017015263.1 |
1238 |
Missense Mutation |
AGG,GGG |
R442G |
XP_016870752.1 |
XM_017015264.1 |
1238 |
Missense Mutation |
AGG,GGG |
R383G |
XP_016870753.1 |
XM_017015265.1 |
1238 |
Missense Mutation |
AGG,GGG |
R349G |
XP_016870754.1 |
XM_017015266.1 |
1238 |
Missense Mutation |
AGG,GGG |
R349G |
XP_016870755.1 |
XM_017015267.1 |
1238 |
Missense Mutation |
AGG,GGG |
R349G |
XP_016870756.1 |
XM_017015268.1 |
1238 |
Missense Mutation |
AGG,GGG |
R349G |
XP_016870757.1 |
- Gene
- SPOUT1
- Gene Name
- SPOUT domain containing methyltransferase 1
There are no transcripts associated with this gene.
View Full Product Details