Product Details

SNP ID
rs147002098
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:112694167 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGATTTGTTGAAGACTGGTTCTGC[A/C]TAAGTAGTTTTCTTTTCTCTTTGTC
Phenotype
MIM: 613273
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
INIP PubMed Links

Gene Details

Gene
INIP
Gene Name
INTS3 and NABP interacting protein
There are no transcripts associated with this gene.

View Full Product Details