Product Details

SNP ID
rs147416046
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34691127 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGAACCAGCAGGCTGAGGGCCA[C/G]TAGCAGGGCCATGGAGGGTGAACAG
Phenotype
MIM: 602227
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CCL19 PubMed Links

Gene Details

Gene
CCL19
Gene Name
C-C motif chemokine ligand 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006274.2 151 Missense Mutation CTG,GTG L5V NP_006265.1

View Full Product Details