Product Details

SNP ID
rs148003612
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35684521 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTCTCCTCCAACAGTTTGATCTC[C/T]TCTTCATATTTATCTTCTTTGGTGG
Phenotype
MIM: 603179 MIM: 190990
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CA9 PubMed Links

Gene Details

Gene
CA9
Gene Name
carbonic anhydrase 9
There are no transcripts associated with this gene.

Gene
TPM2
Gene Name
tropomyosin 2 (beta)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301226.1 908 Silent Mutation GAA,GAG E223E NP_001288155.1
NM_001301227.1 908 Silent Mutation GAA,GAG E223E NP_001288156.1
NM_003289.3 908 Silent Mutation GAA,GAG E223E NP_003280.2
NM_213674.1 908 Silent Mutation GAA,GAG E223E NP_998839.1
XM_017015087.1 908 UTR 3 XP_016870576.1
XM_017015088.1 908 UTR 3 XP_016870577.1
XM_017015089.1 908 Intron XP_016870578.1
XM_017015090.1 908 Silent Mutation GAA,GAG E223E XP_016870579.1
XM_017015091.1 908 Silent Mutation GAA,GAG E223E XP_016870580.1
XM_017015092.1 908 UTR 3 XP_016870581.1
XM_017015093.1 908 UTR 3 XP_016870582.1

View Full Product Details