Product Details

SNP ID
rs148212052
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:68780830 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAGAGATTTTTCCAGGGCATCAC[C/G]AACATGCTTTCTTCTGACGTTGCAC
Phenotype
MIM: 602745
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM122A PubMed Links

Gene Details

Gene
FAM122A
Gene Name
family with sequence similarity 122A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138333.4 825 Silent Mutation ACC,ACG T264T NP_612206.4
Gene
PIP5K1B
Gene Name
phosphatidylinositol-4-phosphate 5-kinase type 1 beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278253.1 825 Intron NP_001265182.1
NM_003558.3 825 Intron NP_003549.1
XM_005252261.3 825 Intron XP_005252318.1
XM_005252262.4 825 Intron XP_005252319.1
XM_006717300.2 825 Intron XP_006717363.1
XM_006717301.1 825 Intron XP_006717364.1
XM_011519082.2 825 Intron XP_011517384.1
XM_011519083.2 825 Intron XP_011517385.1
XM_011519084.2 825 Intron XP_011517386.1
XM_017015188.1 825 Intron XP_016870677.1
XM_017015189.1 825 Intron XP_016870678.1
XM_017015190.1 825 Intron XP_016870679.1
XM_017015191.1 825 Intron XP_016870680.1

View Full Product Details