Product Details
- SNP ID
-
rs148958066
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:132166858 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCTGCATCTGCTGGCGCTCTTCCT[A/G]CACTGCCTCCCTCTGGCCTCTGGGG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NTNG2
PubMed Links
Gene Details
- Gene
- NTNG2
- Gene Name
- netrin G2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032536.2 |
824 |
Silent Mutation |
CTA,CTG |
L9L |
NP_115925.2 |
XM_006717304.3 |
824 |
Silent Mutation |
CTA,CTG |
L9L |
XP_006717367.1 |
XM_011519094.2 |
824 |
Silent Mutation |
CTA,CTG |
L9L |
XP_011517396.1 |
XM_011519096.2 |
824 |
Silent Mutation |
CTA,CTG |
L9L |
XP_011517398.1 |
XM_011519097.2 |
824 |
Silent Mutation |
CTA,CTG |
L9L |
XP_011517399.1 |
XM_011519098.2 |
824 |
Silent Mutation |
CTA,CTG |
L9L |
XP_011517400.1 |
XM_011519099.2 |
824 |
Silent Mutation |
CTA,CTG |
L9L |
XP_011517401.1 |
XM_011519100.2 |
824 |
Silent Mutation |
CTA,CTG |
L9L |
XP_011517402.1 |
XM_011519102.2 |
824 |
Silent Mutation |
CTA,CTG |
L9L |
XP_011517404.1 |
XM_011519103.2 |
824 |
Silent Mutation |
CTA,CTG |
L9L |
XP_011517405.1 |
XM_011519104.2 |
824 |
Silent Mutation |
CTA,CTG |
L9L |
XP_011517406.1 |
XM_011519105.2 |
824 |
Silent Mutation |
CTA,CTG |
L9L |
XP_011517407.1 |
XM_011519106.2 |
824 |
Silent Mutation |
CTA,CTG |
L9L |
XP_011517408.1 |
XM_011519107.2 |
824 |
Silent Mutation |
CTA,CTG |
L9L |
XP_011517409.1 |
XM_011519108.2 |
824 |
Silent Mutation |
CTA,CTG |
L9L |
XP_011517410.1 |
XM_011519109.2 |
824 |
Silent Mutation |
CTA,CTG |
L9L |
XP_011517411.1 |
XM_011519110.2 |
824 |
Silent Mutation |
CTA,CTG |
L9L |
XP_011517412.1 |
XM_011519112.2 |
824 |
Intron |
|
|
XP_011517414.1 |
XM_011519113.2 |
824 |
Intron |
|
|
XP_011517415.1 |
XM_017015212.1 |
824 |
Silent Mutation |
CTA,CTG |
L9L |
XP_016870701.1 |
XM_017015213.1 |
824 |
Silent Mutation |
CTA,CTG |
L9L |
XP_016870702.1 |
XM_017015214.1 |
824 |
Silent Mutation |
CTA,CTG |
L9L |
XP_016870703.1 |
XM_017015215.1 |
824 |
Intron |
|
|
XP_016870704.1 |
XM_017015216.1 |
824 |
Silent Mutation |
CTA,CTG |
L9L |
XP_016870705.1 |
View Full Product Details